ContactUsLinkComponent
Sodium Standard Solutions, (NIST)
Sodium Standard Solutions, (NIST)
Catalog # 104086-104
Supplier:  Hach
CAS Number:  
undefined
Sodium Standard Solutions, (NIST)
Catalog # 104086-104
Supplier:  Hach
Supplier Number:  2318153
CAS Number:  
Specifications
  • Cat. No.:
    104086-104
  • Element:
    Sodium
Specifications
About this item
For accuracy checks and instrument calibrations in sodium determinations.
Caution: 100 mg/L = 100 µg/ml