Order Entry
United States
ContactUsLinkComponent
Potassium Iodide-Iodate Standard Solution, 0.0125 N
Potassium Iodide-Iodate Standard Solution, 0.0125 N
Catalog # 104085-286
Supplier:  Hach
CAS Number:  
undefined
Potassium Iodide-Iodate Standard Solution, 0.0125 N
Catalog # 104085-286
Supplier:  Hach
Supplier Number:  1400153
CAS Number:  

Specifications

  • Description:
    Potassium iodide-iodate standard solution, 0,0125 N
  • Cat. No.:
    104085-286

Specifications

About this item

Used for determination of sulphite by burette iodometric titration.