EDTA Standard Solution
Catalog # 104085-718
Supplier: Hach
CAS Number:
Specifications
- Concentration:5 mg/L as CaCO3
- Description:EDTA standard solution
- Size:4 L
- Cat. No.:104085-718
Specifications
About this item
For hardness determination by burette titration.