Orders LinkContactUsLinkComponent
SINGLET™ pH Buffer, Single-Use
SINGLET™ pH Buffer, Single-Use
Marketsource
Supplier:   Hach
SINGLET™ pH Buffer, Single-Use
Supplier:  Hach
Marketsource
Certificates
SINGLET™ pH Buffer, Single-Use
SINGLET™ pH Buffer, Single-Use
5 Options Available Below
Product Details & Documents
These buffers solutions packs are used for pH meter calibration. The sealed foil pouches can be transported easily and safely without any spillage and also prevents contamination or exposure to air of the solutions.
  • Ideal for on-site analyses
  • Easy to use
  • Accuracy pH: ±0.02 at 25 °C
  • Traceable to NIST
Documents
Product Family Options
Product Information
  • pH accuracy
    Reference temperature
    Packed
    pH
    Availability
    Price
  • ±0.02
    25 °C
    20×20 ml pouches
    4.01
    Log in for availability
    Log in for availability
    Pack of 20
    Pack of 20 - $74.14
    $74.14
    Pack of 20
       $74.14
    Minimum order quantity is 1
    Supplier Number: 2770020
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    25 °C
    20×20 ml pouches
    7
    Log in for availability
    Log in for availability
    Pack of 20
    Pack of 20 - $74.14
    $74.14
    Pack of 20
       $74.14
    Minimum order quantity is 1
    Supplier Number: 2770120
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    25 °C
    20×20 ml pouches
    10.01
    Log in for availability
    Log in for availability
    Pack of 20
    Pack of 20 - $74.14
    $74.14
    Pack of 20
       $74.14
    Minimum order quantity is 1
    Supplier Number: 2770220
    Specifications:
    Marketsource
    Marketsource
Combination packs
Product Information
  • pH accuracy
    Reference temperature
    Packed
    pH
    Availability
    Price
  • 20/pack (10 each of 4.01 and 7.00 pH)
    4.01/7.00
    Log in for availability
    Log in for availability
    Each
    Each - $72.95
    $72.95
    Each
       $72.95
    Minimum order quantity is 1
    Supplier Number: 2769920
    Specifications:
    Marketsource
    Marketsource
  • 20/pack (10 each of 7.00 and 10.01 pH)
    7.00/10.01
    Log in for availability
    Log in for availability
    Each
    Each - $72.95
    $72.95
    Each
       $72.95
    Minimum order quantity is 1
    Supplier Number: 2769820
    Specifications:
    Marketsource
    Marketsource