Order Entry
United States
Orders LinkContactUsLinkComponent
pH Buffer Solutions, Color Coded

pH Buffer Solutions, Color Coded

Marketsource
Supplier:   Hach
pH Buffer Solutions, Color Coded
Supplier:  Hach
Marketsource
Certificates
pH Buffer Solutions, Color Coded
pH Buffer Solutions, Color Coded

12 Options Available Below

Product Details & Documents
  • For calibration of pH meters
  • Color coded for easy identification
  • Accuracy: pH is ±0.02 at 25 °C

Certifications: Traceable to NIST.

Documents

Product Family Options

Individual pH buffer solutions

Product Information

  • pH accuracy
    pH buffer type
    Reference temperature
    pH
    Availability
    Price
  • ±0.02
    Phthalate
    25 °C
    4.01
    Log in for availability
    Log in for availability
    Each
    Each - $12.45
    $12.45
    Each
       $12.45
    Minimum order quantity is 1

    Supplier Number: 2283436
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Phthalate
    25 °C
    4.01
    Log in for availability
    Log in for availability
    Each
    Each - $19.09
    $19.09
    Each
       $19.09
    Minimum order quantity is 1

    Supplier Number: 2283426
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Phthalate
    25 °C
    4.01
    Log in for availability
    Log in for availability
    Each
    Each - $108.83
    $108.83
    Each
       $108.83
    Minimum order quantity is 1

    Supplier Number: 2283456
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Phthalate
    25 °C
    4.01
    Log in for availability
    Log in for availability
    Each
    Each - $300.46
    $300.46
    Each
       $300.46
    Minimum order quantity is 1

    Supplier Number: 2283461
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Phosphate
    25 °C
    7
    Log in for availability
    Log in for availability
    Each
    Each - $12.45
    $12.45
    Each
       $12.45
    Minimum order quantity is 1

    Supplier Number: 2283536
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Phosphate
    25 °C
    7
    Log in for availability
    Log in for availability
    Each
    Each - $19.09
    $19.09
    Each
       $19.09
    Minimum order quantity is 1

    Supplier Number: 2283526
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Phosphate
    25 °C
    7
    Log in for availability
    Log in for availability
    Each
    Each - $108.83
    $108.83
    Each
       $108.83
    Minimum order quantity is 1

    Supplier Number: 2283556
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Phosphate
    25 °C
    7
    Log in for availability
    Log in for availability
    Each
    Each - $300.46
    $300.46
    Each
       $300.46
    Minimum order quantity is 1

    Supplier Number: 2283561
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Carbonate
    25 °C
    10.01
    Log in for availability
    Log in for availability
    Each
    Each - $19.09
    $19.09
    Each
       $19.09
    Minimum order quantity is 1

    Supplier Number: 2283626
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Carbonate
    25 °C
    10.01
    Log in for availability
    Log in for availability
    Each
    Each - $108.83
    $108.83
    Each
       $108.83
    Minimum order quantity is 1

    Supplier Number: 2283656
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Carbonate
    25 °C
    10.01
    Log in for availability
    Log in for availability
    Each
    Each - $300.46
    $300.46
    Each
       $300.46
    Minimum order quantity is 1

    Supplier Number: 2283661
    Specifications:
    Marketsource
    Marketsource

Buffer kit

Product Information

  • pH accuracy
    pH buffer type
    Reference temperature
    pH
    Availability
    Price
  • -
    4.01/7.00/10.01
    Log in for availability
    Log in for availability
    Each
    Each - $58.55
    $58.55
    Each
       $58.55
    Minimum order quantity is 1

    Supplier Number: 2947600
    Specifications:
    Marketsource
    Marketsource