sensION+ 5056 ORP Electrode
104087-702
: Hach
- Cat. No.:104087-702
- For use with:General aqueous applications
- Junction:Ceramic pin
- mV Range:±2000
- Connector:BNC, 1 m cable
- Electrolyte:Gel (non refillable)
sensION+ 5056 is a combination platinum ORP electrode with a glass body and non refillable gel-electrolyte reference. It is dedicated for use with Hach sensION+ labpratory pH (mV) meters.
- Fixed 1 meter cable and BNC connector
- Low-maintenance design for a variety of applications
- Gel-electrolyte reference and platinum wire sensor provides accurate, stable results over a wide range of applications
- Encapsulated cartridge reference system ensures premium stability and long lifespan
- Innovative reference system utilises a “cartridge” of Ag/AgCl crystals contacting a silver wire inside a glass tube
The 5056 is ideal for oxidation-reduction potential (ORP) measurements in general aqueous applications.
: Supplied with basic user manual.