Combined pH Electrode, Red Rod
Type 7 plug, 1 m cable, Saturated KCl (refillable), 0 - 12, Porous pin
104085-058
Marketsource
: Hach
Supplier #: 945-252
The Radiometer GK2401C combination pH electrode has a glass body, Red-Rod reference system, refillable electrolyte (saturated KCl) and Radiometer Type 7 connector for compatibility with Radiometer MeterLab legacy meters.
- Red-Rod electrodes provide a stable signal fast by protecting the reference elements from light interference
- The Red-Rod saturated KCl salt-bridge also aids in creating a stable liquid junction potential
- Red-Rod system creates a trouble-free liquid junction: No silver ions and therefore less risk of clogging when measuring in challenging sample matrices
- Red-Rod reference system enables use over a wider range of temperatures than for most pH electrodes
- pH range:0 - 12
- Junction:Porous pin
- Connector:Type 7 plug, 1 m cable
- Electrolyte:Saturated KCl (refillable)