Radiometer Analytical pH Electrode
General purposes, Screw cap (no cable), 0 - 12
104087-210
Marketsource
: Hach
: E11M001
These glass electrodes are compatible with Hach sensION+ benchtop pH meters (or any meter with a BNC connection). The PHG301 pH electrode should be used with Ag/AgCl or calomel reference electrodes.
- Made with highly shock-resistant glass, making them extremely tough
- Average lifetime of a correctly maintained Radiometer glass pH electrode is approximately two years
The PHG301 is a screw cap electrode and requires the CL114 cable for connection to a meter.
: Supplied with certificate of conformity.
- For use with:General purposes
- pH range:0 - 12
- Connector:Screw cap (no cable)



