sensION+ 5059 Multi-Parameter Electrode
Catalog # 104087-706
Supplier: Hach
Specifications
- Cat. No.:104087-706
- Measurement range:0 - 14 pH
- For use with:General aqueous applications
- Junction:Ceramic pin
- Fill solution:Gel (non refillable)
- Connector:Sension+ MP8, fixed 1 m cable
- Conductivity:20 µS/cm - 200 mS/cm
Specifications
About this item
sensION+ 5059 is a combination multi-parameter electrode for determination of pH, conductivity and temperature. It has a plastic body, non-refillable gel reference electrolyte and built-in temperature sensor.
- Determine multiple parameters in the field with one standard sized electrode
- Low maintenance design for a variety of applications
- Ultra portable but protected against harsh field conditions
- Heavy-duty electrode handle design optimised for field calibration and storage
- Glass (pH); platinum (conductivity); and Pt1000 (temperature) sensors
Dedicated for use with Hach sensION+ MM150 portable meters. Ideal for pH, conductivity and temperature measurements in general aqueous applications.
Delivery: Supplied with basic user manual.