Order Entry
United States
ContactUsLinkComponent
sensION+ 5048 Multi-Parameter Electrode (pH/ORP/Conductivity)
sensION+ 5048 Multi-Parameter Electrode (pH/ORP/Conductivity)
Catalog # 104087-692
Supplier:  Hach
undefined
sensION+ 5048 Multi-Parameter Electrode (pH/ORP/Conductivity)
Catalog # 104087-692
Supplier:  Hach
Supplier Number:  LZW5048970002

Specifications

  • Cat. No.:
    104087-692
  • Measurement range:
    0 - 14 pH
  • For use with:
    General aqueous applications
  • Junction:
    Ceramic pin
  • Fill solution:
    Gel (non refillable)
  • Connector:
    MP8, 1.5 m cable
  • Conductivity:
    20 µS/cm - 200 mS/cm

Specifications

About this item

Multisensor for pH, ORP, conductivity and temperature. For use with sensION+ MM150 multi-parameter meter.

  • Accurate
  • Easy to use
  • Rugged and durable
  • Automatic temperature compensation
  • Glass (pH); platinum (conductivity & ORP); and Pt1000 (temperature) sensors

Attachable calibration tubes can screw into handle of electrode (only 2 ml of buffer or standard). Storage protector prevents breakage and avoids over pressurisation of the diaphragm. Ideal for pH, conductivity, ORP and temperature measurements in general aqueous applications.