Order Entry
United States
ContactUsLinkComponent
 
sensION+ 5045 Multi-Parameter Electrode (pH/ORP/Temperature)
  104087-690
 :  Hach
undefined
sensION+ 5045 Multi-Parameter Electrode (pH/ORP/Temperature)
  104087-690
 :  Hach
 :  LZW5045970002

 

  • Cat. No.:
    104087-690
  • Measurement range:
    0 - 14 pH
  • For use with:
    General aqueous applications
  • Junction:
    Ceramic pin
  • Fill solution:
    Gel (non refillable)
  • Connector:
    MP8, 1.5 m cable

 

 

sensION+ 5045 is a combination multi-parameter electrode for determination of pH, ORP and temperature. Dedicated for use with Hach sensION+ MM110 portable meters.

  • Ceramic pin reference junction and glass (pH); platinum wire (ORP); and Pt1000 (temperature) sensors
  • Plastic body, non refillable gel reference electrolyte and built-in temperature sensor
  • Low maintenance design for a variety of applications
  • Ultra portable but protected against harsh field conditions
  • Heavy-duty electrode handle design optimised for field calibration and storage
  • Innovative designs house multiple sensing elements in a standard 12 mm electrode body

The electrode is ideal for pH, ORP and temperature measurements in general aqueous applications.