ContactUsLinkComponent
sensION+ 9655 Ion Selective Electrode, Half-Cell, Fluoride
sensION+ 9655 Ion Selective Electrode, Half-Cell, Fluoride
Catalog # 104087-722
Supplier:  Hach
undefined
sensION+ 9655 Ion Selective Electrode, Half-Cell, Fluoride
Catalog # 104087-722
Supplier:  Hach
Supplier Number:  LZW9655970002
Specifications
  • Cat. No.:
    104087-722
  • Fill solution:
    Fluoride
  • Connector:
    BNC, fixed 1 m cable
Specifications
About this item
sensION+ 9655 is a half-cell (reference not integrated) fluoride ion selective electrode with an epoxy body and non-replaceable crystalline membrane selective to fluoride ions in solution.
  • Requires virtually no maintenance: Solid-state sensor design allows for dry storage of the ISE without a shelf life or membrane replacement
  • Provides fast, stable, and accurate response in a variety of sample types
  • Recommended for use with the sensION+ 5044 reference electrode
The 9655 has a fixed 1 meter cable and BNC connector for laboratory use with the Hach sensION+ MM374 Multimeter. The electrode is ideal for measuring fluoride concentrations in drinking water, wastewater and general water quality applications.
Delivery: Supplied with basic user manual.