Order Entry
United States
Orders LinkContactUsLinkComponent
Multi-Parameter Meter, Bench, HQ440d

Multi-Parameter Meter, Bench, HQ440d

Marketsource
Supplier:   Hach
Multi-Parameter Meter, Bench, HQ440d
Supplier:  Hach
Marketsource
Certificates
Multi-Parameter Meter, Bench, HQ440d

7 Options Available Below

Product Details & Documents

The Hach HQ440d benchtop meter is an advanced laboratory meter designed for water quality experts. It features two input channels for simultaneous measurement of pH, conductivity, DO, BOD, ORP, Ammonia, Ammonium, Fluoride, Chloride, Sodium, and temperature − with any IntelliCAL™ smart probe.

  • Intuitive user interface for simple operation and accurate results
  • Automatic probe and parameter recognition
  • IntelliCAL™ smart probes store calibration data in the probe
  • GLP compatible: HQd smart system records serial numbers, current calibration data, user ID, sample ID, time, and date automatically
  • Ultra-bright screen and large font size for easy to read results
  • Internal USB ports simplify data transfer, peripherals, and software updates and connection to PC, printer and keyboards

HQd meters connect with smart probes that automatically recognise the testing parameter, calibration history, and method settings to minimize errors and setup time. The benchtop meter is designed to withstand years of continuous lab use. HQd meters feature a simple user interface that does not require manuals or training to operate.

Suitable applications for the electrodes:
PHC705 + PHC805: General purposes
PHC705A: Alkaline samples
PHC725: Drinking water, low ionic strength (LIS) solutions
PHC729: Surface measurements
PHC735: 'Dirty' samples, wastewater, food and beverage applications
PHC745: 'challenging' samples, wastewater influent and other solutions prone to clogging traditional junctions
PHC108: Semi-solid samples (sludge, meat and cheese)
PHC281: Difficult samples (low ionic strength (LIS), ultrapure, dirty, TRIS, or high-solid content samples)

Delivery information: All meters are supplied with electrode, pH buffer kit (500 ml), pH storage solution (500 ml), probe stand, 4 AA batteries, power adapter, USB cable for data transfer, quick-start guide, user manual, and documentation CD.

Specifications for Product Family

Specifications

  • Catalog No:
  • 104086-938
  • 104086-936
  • 104086-940
  • 104086-934
  • 104086-944
  • 104086-932
  • 104086-930
  • pH Range:
  • 0 - 14
  • 0 - 14
  • 0 - 14
  • 0 - 14
  • 0 - 14
  • 0 - 14
  • 0 - 14
  • pH Resolution:
  • 0.001 - 0.1 (selectable)
  • 0.001 - 0.1 (selectable)
  • 0.001 - 0.1 (selectable)
  • 0.001 - 0.1 (selectable)
  • 0.001 - 0.1 (selectable)
  • 0.001 - 0.1 (selectable)
  • 0.001 - 0.1 (selectable)
  • pH Accuracy:
  • ±0.002
  • ±0.002
  • ±0.002
  • ±0.002
  • ±0.002
  • ±0.002
  • ±0.002
  • pH Calibration:
  • 4-point calibration
  • 4-point calibration
  • 4-point calibration
  • 4-point calibration
  • 4-point calibration
  • 4-point calibration
  • 4-point calibration
  • mV range:
  • ±1500
  • ±1500
  • ±1500
  • ±1500
  • ±1500
  • ±1500
  • ±1500
  • mV resolution:
  • 0.1
  • 0.1
  • 0.1
  • 0.1
  • 0.1
  • 0.1
  • 0.1
  • mV accuracy:
  • ±0.1
  • ±0.1
  • ±0.1
  • ±0.1
  • ±0.1
  • ±0.1
  • ±0.1
  • Conductivity Range:
  • 0.01 μS/cm - 200 mS/cm
  • 0.01 μS/cm - 200 mS/cm
  • 0.01 μS/cm - 200 mS/cm
  • 0.01 μS/cm - 200 mS/cm
  • 0.01 μS/cm - 200 mS/cm
  • 0.01 μS/cm - 200 mS/cm
  • 0.01 μS/cm - 200 mS/cm
  • Conductivity Resolution:
  • 5 digits with 2 digits after decimal point
  • 5 digits with 2 digits after decimal point
  • 5 digits with 2 digits after decimal point
  • 5 digits with 2 digits after decimal point
  • 5 digits with 2 digits after decimal point
  • 5 digits with 2 digits after decimal point
  • 5 digits with 2 digits after decimal point
  • Conductivity Accuracy:
  • ±0.5% (1 µS/cm - 200 mS/cm)
  • ±0.5% (1 µS/cm - 200 mS/cm)
  • ±0.5% (1 µS/cm - 200 mS/cm)
  • ±0.5% (1 µS/cm - 200 mS/cm)
  • ±0.5% (1 µS/cm - 200 mS/cm)
  • ±0.5% (1 µS/cm - 200 mS/cm)
  • ±0.5% (1 µS/cm - 200 mS/cm)
  • Resistivity range:
  • 2.5 Ωcm - 49 MΩcm
  • 2.5 Ωcm - 49 MΩcm
  • 2.5 Ωcm - 49 MΩcm
  • 2.5 Ωcm - 49 MΩcm
  • 2.5 Ωcm - 49 MΩcm
  • 2.5 Ωcm - 49 MΩcm
  • 2.5 Ωcm - 49 MΩcm
  • Resistivity accuracy:
  • 0.005
  • 0.005
  • 0.005
  • 0.005
  • 0.005
  • 0.005
  • 0.005
  • Salinity Range:
  • 0 - 42 g/kg
  • 0 - 42 g/kg
  • 0 - 42 g/kg
  • 0 - 42 g/kg
  • 0 - 42 g/kg
  • 0 - 42 g/kg
  • 0 - 42 g/kg
  • Salinity Resolution:
  • 0.01 ppt
  • 0.01 ppt
  • 0.01 ppt
  • 0.01 ppt
  • 0.01 ppt
  • 0.01 ppt
  • 0.01 ppt
  • Salinity Accuracy:
  • 0.01 ppt
  • 0.01 ppt
  • 0.01 ppt
  • 0.01 ppt
  • 0.01 ppt
  • 0.01 ppt
  • 0.01 ppt
  • TDS Range:
  • 0.0 - 50.0 mg/L
  • 0.0 - 50.0 mg/L
  • 0.0 - 50.0 mg/L
  • 0.0 - 50.0 mg/L
  • 0.0 - 50.0 mg/L
  • 0.0 - 50.0 mg/L
  • 0.0 - 50.0 mg/L
  • TDS Resolution:
  • 0.1/ 0.01/ 0.001
  • 0.1/ 0.01/ 0.001
  • 0.1/ 0.01/ 0.001
  • 0.1/ 0.01/ 0.001
  • 0.1/ 0.01/ 0.001
  • 0.1/ 0.01/ 0.001
  • 0.1/ 0.01/ 0.001
  • DO Concentration:
  • 0.01 - 20 mg/L
  • 0.01 - 20 mg/L
  • 0.01 - 20 mg/L
  • 0.01 - 20 mg/L
  • 0.01 - 20 mg/L
  • 0.01 - 20 mg/L
  • 0.01 - 20 mg/L
  • DO Resolution:
  • 0.01 mg/L
  • 0.01 mg/L
  • 0.01 mg/L
  • 0.01 mg/L
  • 0.01 mg/L
  • 0.01 mg/L
  • 0.01 mg/L
  • DO Saturation:
  • 0 - 200 %
  • 0 - 200 %
  • 0 - 200 %
  • 0 - 200 %
  • 0 - 200 %
  • 0 - 200 %
  • 0 - 200 %
  • Temperature Resolution:
  • 0.1 °C
  • 0.1 °C
  • 0.1 °C
  • 0.1 °C
  • 0.1 °C
  • 0.1 °C
  • 0.1 °C
  • Temperature Accuracy:
  • ± 0.3 °C
  • ± 0.3 °C
  • ± 0.3 °C
  • ± 0.3 °C
  • ± 0.3 °C
  • ± 0.3 °C
  • ± 0.3 °C
  • Temperature Compensation:
  • Off, automatic and manual (parameter dependent)
  • Off, automatic and manual (parameter dependent)
  • Off, automatic and manual (parameter dependent)
  • Off, automatic and manual (parameter dependent)
  • Off, automatic and manual (parameter dependent)
  • Off, automatic and manual (parameter dependent)
  • Off, automatic and manual (parameter dependent)
  • Display:
  • LCD (240×160 pixels) with backlight
  • LCD (240×160 pixels) with backlight
  • LCD (240×160 pixels) with backlight
  • LCD (240×160 pixels) with backlight
  • LCD (240×160 pixels) with backlight
  • LCD (240×160 pixels) with backlight
  • LCD (240×160 pixels) with backlight
  • Power supply:
  • Power adapter (100 - 240 VAC, 50/60 Hz) or 4 × AA Alkaline or rechargeable Nickel Metal Hydride (NiMH) batteries
  • Power adapter (100 - 240 VAC, 50/60 Hz) or 4 × AA Alkaline or rechargeable Nickel Metal Hydride (NiMH) batteries
  • Power adapter (100 - 240 VAC, 50/60 Hz) or 4 × AA Alkaline or rechargeable Nickel Metal Hydride (NiMH) batteries
  • Power adapter (100 - 240 VAC, 50/60 Hz) or 4 × AA Alkaline or rechargeable Nickel Metal Hydride (NiMH) batteries
  • Power adapter (100 - 240 VAC, 50/60 Hz) or 4 × AA Alkaline or rechargeable Nickel Metal Hydride (NiMH) batteries
  • Power adapter (100 - 240 VAC, 50/60 Hz) or 4 × AA Alkaline or rechargeable Nickel Metal Hydride (NiMH) batteries
  • Power adapter (100 - 240 VAC, 50/60 Hz) or 4 × AA Alkaline or rechargeable Nickel Metal Hydride (NiMH) batteries
  • Compliance:
  • IP 67, IP 54, GLP
  • IP 67, IP 54, GLP
  • IP 67, IP 54, GLP
  • IP 67, IP 54, GLP
  • IP 67, IP 54, GLP
  • IP 67, IP 54, GLP
  • IP 67, IP 54, GLP
  • Data Logging - Log Function:
  • Manual or automatic
  • Manual or automatic
  • Manual or automatic
  • Manual or automatic
  • Manual or automatic
  • Manual or automatic
  • Manual or automatic
  • Data Logging Capacity:
  • 500 data results
  • 500 data results
  • 500 data results
  • 500 data results
  • 500 data results
  • 500 data results
  • 500 data results
  • Outputs:
  • USB
  • USB
  • USB
  • USB
  • USB
  • USB
  • USB
  • Input:
  • 2 × M12 Verbindung für IntelliCAL™ Sensoren
  • 2 × M12 Verbindung für IntelliCAL™ Sensoren
  • 2 × M12 Verbindung für IntelliCAL™ Sensoren
  • 2 × M12 Verbindung für IntelliCAL™ Sensoren
  • 2 × M12 Verbindung für IntelliCAL™ Sensoren
  • 2 × M12 Verbindung für IntelliCAL™ Sensoren
  • 2 × M12 Verbindung für IntelliCAL™ Sensoren
  • W×D×H:
  • 173×234×89 mm
  • 173×234×89 mm
  • 173×234×89 mm
  • 173×234×89 mm
  • 173×234×89 mm
  • 173×234×89 mm
  • 173×234×89 mm
  • Weight:
  • 0.85 kg
  • 0.85 kg
  • 0.85 kg
  • 0.85 kg
  • 0.85 kg
  • 0.85 kg
  • 0.85 kg

Specifications

Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    Each
    Each - $5,498.31
    $5,498.31
    Each
       $5,498.31
    Minimum order quantity is 1

    Supplier Number: 8506250
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    104086-938
    pH Range
    0 - 14
    pH Resolution
    0.001 - 0.1 (selectable)
    pH Accuracy
    ±0.002
    pH Calibration
    4-point calibration
    mV range
    ±1500
    mV resolution
    0.1
    mV accuracy
    ±0.1
    Conductivity Range
    0.01 μS/cm - 200 mS/cm
    Conductivity Resolution
    5 digits with 2 digits after decimal point
    Conductivity Accuracy
    ±0.5% (1 µS/cm - 200 mS/cm)
    Resistivity range
    2.5 Ωcm - 49 MΩcm
    Resistivity accuracy
    0.005
    Salinity Range
    0 - 42 g/kg
    Salinity Resolution
    0.01 ppt
    Salinity Accuracy
    0.01 ppt
    TDS Range
    0.0 - 50.0 mg/L
    TDS Resolution
    0.1/ 0.01/ 0.001
    DO Concentration
    0.01 - 20 mg/L
    DO Resolution
    0.01 mg/L
    DO Saturation
    0 - 200 %
    Temperature Resolution
    0.1 °C
    Temperature Accuracy
    ± 0.3 °C
    Temperature Compensation
    Off, automatic and manual (parameter dependent)
    Display
    LCD (240×160 pixels) with backlight
    Power supply
    Power adapter (100 - 240 VAC, 50/60 Hz) or 4 × AA Alkaline or rechargeable Nickel Metal Hydride (NiMH) batteries
    Compliance
    IP 67, IP 54, GLP
    Data Logging - Log Function
    Manual or automatic
    Data Logging Capacity
    500 data results
    Outputs
    USB
    Input
    2 × M12 Verbindung für IntelliCAL™ Sensoren
    W×D×H
    173×234×89 mm
    Weight
    0.85 kg
    Marketsource
  • Log in for availability
    Log in for availability
    Each
    Each - $5,631.23
    $5,631.23
    Each
       $5,631.23
    Minimum order quantity is 1

    Supplier Number: 8506050
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    104086-936
    pH Range
    0 - 14
    pH Resolution
    0.001 - 0.1 (selectable)
    pH Accuracy
    ±0.002
    pH Calibration
    4-point calibration
    mV range
    ±1500
    mV resolution
    0.1
    mV accuracy
    ±0.1
    Conductivity Range
    0.01 μS/cm - 200 mS/cm
    Conductivity Resolution
    5 digits with 2 digits after decimal point
    Conductivity Accuracy
    ±0.5% (1 µS/cm - 200 mS/cm)
    Resistivity range
    2.5 Ωcm - 49 MΩcm
    Resistivity accuracy
    0.005
    Salinity Range
    0 - 42 g/kg
    Salinity Resolution
    0.01 ppt
    Salinity Accuracy
    0.01 ppt
    TDS Range
    0.0 - 50.0 mg/L
    TDS Resolution
    0.1/ 0.01/ 0.001
    DO Concentration
    0.01 - 20 mg/L
    DO Resolution
    0.01 mg/L
    DO Saturation
    0 - 200 %
    Temperature Resolution
    0.1 °C
    Temperature Accuracy
    ± 0.3 °C
    Temperature Compensation
    Off, automatic and manual (parameter dependent)
    Display
    LCD (240×160 pixels) with backlight
    Power supply
    Power adapter (100 - 240 VAC, 50/60 Hz) or 4 × AA Alkaline or rechargeable Nickel Metal Hydride (NiMH) batteries
    Compliance
    IP 67, IP 54, GLP
    Data Logging - Log Function
    Manual or automatic
    Data Logging Capacity
    500 data results
    Outputs
    USB
    Input
    2 × M12 Verbindung für IntelliCAL™ Sensoren
    W×D×H
    173×234×89 mm
    Weight
    0.85 kg
    Marketsource
  • Log in for availability
    Log in for availability
    Each
    Each - $5,631.23
    $5,631.23
    Each
       $5,631.23
    Minimum order quantity is 1

    Supplier Number: 8506450
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    104086-940
    pH Range
    0 - 14
    pH Resolution
    0.001 - 0.1 (selectable)
    pH Accuracy
    ±0.002
    pH Calibration
    4-point calibration
    mV range
    ±1500
    mV resolution
    0.1
    mV accuracy
    ±0.1
    Conductivity Range
    0.01 μS/cm - 200 mS/cm
    Conductivity Resolution
    5 digits with 2 digits after decimal point
    Conductivity Accuracy
    ±0.5% (1 µS/cm - 200 mS/cm)
    Resistivity range
    2.5 Ωcm - 49 MΩcm
    Resistivity accuracy
    0.005
    Salinity Range
    0 - 42 g/kg
    Salinity Resolution
    0.01 ppt
    Salinity Accuracy
    0.01 ppt
    TDS Range
    0.0 - 50.0 mg/L
    TDS Resolution
    0.1/ 0.01/ 0.001
    DO Concentration
    0.01 - 20 mg/L
    DO Resolution
    0.01 mg/L
    DO Saturation
    0 - 200 %
    Temperature Resolution
    0.1 °C
    Temperature Accuracy
    ± 0.3 °C
    Temperature Compensation
    Off, automatic and manual (parameter dependent)
    Display
    LCD (240×160 pixels) with backlight
    Power supply
    Power adapter (100 - 240 VAC, 50/60 Hz) or 4 × AA Alkaline or rechargeable Nickel Metal Hydride (NiMH) batteries
    Compliance
    IP 67, IP 54, GLP
    Data Logging - Log Function
    Manual or automatic
    Data Logging Capacity
    500 data results
    Outputs
    USB
    Input
    2 × M12 Verbindung für IntelliCAL™ Sensoren
    W×D×H
    173×234×89 mm
    Weight
    0.85 kg
    Marketsource
  • Log in for availability
    Log in for availability
    Each
    Each - $5,631.23
    $5,631.23
    Each
       $5,631.23
    Minimum order quantity is 1

    Supplier Number: 8505850
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    104086-934
    pH Range
    0 - 14
    pH Resolution
    0.001 - 0.1 (selectable)
    pH Accuracy
    ±0.002
    pH Calibration
    4-point calibration
    mV range
    ±1500
    mV resolution
    0.1
    mV accuracy
    ±0.1
    Conductivity Range
    0.01 μS/cm - 200 mS/cm
    Conductivity Resolution
    5 digits with 2 digits after decimal point
    Conductivity Accuracy
    ±0.5% (1 µS/cm - 200 mS/cm)
    Resistivity range
    2.5 Ωcm - 49 MΩcm
    Resistivity accuracy
    0.005
    Salinity Range
    0 - 42 g/kg
    Salinity Resolution
    0.01 ppt
    Salinity Accuracy
    0.01 ppt
    TDS Range
    0.0 - 50.0 mg/L
    TDS Resolution
    0.1/ 0.01/ 0.001
    DO Concentration
    0.01 - 20 mg/L
    DO Resolution
    0.01 mg/L
    DO Saturation
    0 - 200 %
    Temperature Resolution
    0.1 °C
    Temperature Accuracy
    ± 0.3 °C
    Temperature Compensation
    Off, automatic and manual (parameter dependent)
    Display
    LCD (240×160 pixels) with backlight
    Power supply
    Power adapter (100 - 240 VAC, 50/60 Hz) or 4 × AA Alkaline or rechargeable Nickel Metal Hydride (NiMH) batteries
    Compliance
    IP 67, IP 54, GLP
    Data Logging - Log Function
    Manual or automatic
    Data Logging Capacity
    500 data results
    Outputs
    USB
    Input
    2 × M12 Verbindung für IntelliCAL™ Sensoren
    W×D×H
    173×234×89 mm
    Weight
    0.85 kg
    Marketsource
  • Log in for availability
    Log in for availability
    Each
    Each - $5,887.38
    $5,887.38
    Each
       $5,887.38
    Minimum order quantity is 1

    Supplier Number: 8507450
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    104086-944
    pH Range
    0 - 14
    pH Resolution
    0.001 - 0.1 (selectable)
    pH Accuracy
    ±0.002
    pH Calibration
    4-point calibration
    mV range
    ±1500
    mV resolution
    0.1
    mV accuracy
    ±0.1
    Conductivity Range
    0.01 μS/cm - 200 mS/cm
    Conductivity Resolution
    5 digits with 2 digits after decimal point
    Conductivity Accuracy
    ±0.5% (1 µS/cm - 200 mS/cm)
    Resistivity range
    2.5 Ωcm - 49 MΩcm
    Resistivity accuracy
    0.005
    Salinity Range
    0 - 42 g/kg
    Salinity Resolution
    0.01 ppt
    Salinity Accuracy
    0.01 ppt
    TDS Range
    0.0 - 50.0 mg/L
    TDS Resolution
    0.1/ 0.01/ 0.001
    DO Concentration
    0.01 - 20 mg/L
    DO Resolution
    0.01 mg/L
    DO Saturation
    0 - 200 %
    Temperature Resolution
    0.1 °C
    Temperature Accuracy
    ± 0.3 °C
    Temperature Compensation
    Off, automatic and manual (parameter dependent)
    Display
    LCD (240×160 pixels) with backlight
    Power supply
    Power adapter (100 - 240 VAC, 50/60 Hz) or 4 × AA Alkaline or rechargeable Nickel Metal Hydride (NiMH) batteries
    Compliance
    IP 67, IP 54, GLP
    Data Logging - Log Function
    Manual or automatic
    Data Logging Capacity
    500 data results
    Outputs
    USB
    Input
    2 × M12 Verbindung für IntelliCAL™ Sensoren
    W×D×H
    173×234×89 mm
    Weight
    0.85 kg
    Marketsource
  • Log in for availability
    Log in for availability
    Each
    Each - $5,758.62
    $5,758.62
    Each
       $5,758.62
    Minimum order quantity is 1

    Supplier Number: 8505450
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    104086-932
    pH Range
    0 - 14
    pH Resolution
    0.001 - 0.1 (selectable)
    pH Accuracy
    ±0.002
    pH Calibration
    4-point calibration
    mV range
    ±1500
    mV resolution
    0.1
    mV accuracy
    ±0.1
    Conductivity Range
    0.01 μS/cm - 200 mS/cm
    Conductivity Resolution
    5 digits with 2 digits after decimal point
    Conductivity Accuracy
    ±0.5% (1 µS/cm - 200 mS/cm)
    Resistivity range
    2.5 Ωcm - 49 MΩcm
    Resistivity accuracy
    0.005
    Salinity Range
    0 - 42 g/kg
    Salinity Resolution
    0.01 ppt
    Salinity Accuracy
    0.01 ppt
    TDS Range
    0.0 - 50.0 mg/L
    TDS Resolution
    0.1/ 0.01/ 0.001
    DO Concentration
    0.01 - 20 mg/L
    DO Resolution
    0.01 mg/L
    DO Saturation
    0 - 200 %
    Temperature Resolution
    0.1 °C
    Temperature Accuracy
    ± 0.3 °C
    Temperature Compensation
    Off, automatic and manual (parameter dependent)
    Display
    LCD (240×160 pixels) with backlight
    Power supply
    Power adapter (100 - 240 VAC, 50/60 Hz) or 4 × AA Alkaline or rechargeable Nickel Metal Hydride (NiMH) batteries
    Compliance
    IP 67, IP 54, GLP
    Data Logging - Log Function
    Manual or automatic
    Data Logging Capacity
    500 data results
    Outputs
    USB
    Input
    2 × M12 Verbindung für IntelliCAL™ Sensoren
    W×D×H
    173×234×89 mm
    Weight
    0.85 kg
    Marketsource
  • Log in for availability
    Log in for availability
    Each
    Each - $5,267.08
    $5,267.08
    Each
       $5,267.08
    Minimum order quantity is 1

    Supplier Number: 8505350
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    104086-930
    pH Range
    0 - 14
    pH Resolution
    0.001 - 0.1 (selectable)
    pH Accuracy
    ±0.002
    pH Calibration
    4-point calibration
    mV range
    ±1500
    mV resolution
    0.1
    mV accuracy
    ±0.1
    Conductivity Range
    0.01 μS/cm - 200 mS/cm
    Conductivity Resolution
    5 digits with 2 digits after decimal point
    Conductivity Accuracy
    ±0.5% (1 µS/cm - 200 mS/cm)
    Resistivity range
    2.5 Ωcm - 49 MΩcm
    Resistivity accuracy
    0.005
    Salinity Range
    0 - 42 g/kg
    Salinity Resolution
    0.01 ppt
    Salinity Accuracy
    0.01 ppt
    TDS Range
    0.0 - 50.0 mg/L
    TDS Resolution
    0.1/ 0.01/ 0.001
    DO Concentration
    0.01 - 20 mg/L
    DO Resolution
    0.01 mg/L
    DO Saturation
    0 - 200 %
    Temperature Resolution
    0.1 °C
    Temperature Accuracy
    ± 0.3 °C
    Temperature Compensation
    Off, automatic and manual (parameter dependent)
    Display
    LCD (240×160 pixels) with backlight
    Power supply
    Power adapter (100 - 240 VAC, 50/60 Hz) or 4 × AA Alkaline or rechargeable Nickel Metal Hydride (NiMH) batteries
    Compliance
    IP 67, IP 54, GLP
    Data Logging - Log Function
    Manual or automatic
    Data Logging Capacity
    500 data results
    Outputs
    USB
    Input
    2 × M12 Verbindung für IntelliCAL™ Sensoren
    W×D×H
    173×234×89 mm
    Weight
    0.85 kg
    Marketsource