ContactUsLinkComponent
Separating Funnels, Squibb, HDPE Stopper, PTFE Stopcock
Separating Funnels, Squibb, HDPE Stopper, PTFE Stopcock
Catalog # 104086-876
Supplier:  Hach
undefined
Separating Funnels, Squibb, HDPE Stopper, PTFE Stopcock
Catalog # 104086-876
Supplier:  Hach
Supplier Number:  52049
Specifications
  • Capacity:
    500 ml
  • Cat. No.:
    104086-876
Specifications
About this item
Pear-shaped (Squibb) glass separatory funnel with PTFE stopcock and HDPE stopper.
  • Clear glass body allows for easy detection of layers between immiscible liquids
  • Highly chemical resistant and fully autoclavable
  • HDPE stopper with closed bottom