Block Rack D12
About this item
High-quality, safe, and robust racks and boxes are essential for specimen processing and storage. SARSTEDT offers the right product for every laboratory and analytical requirement.
- Format: 5×10 configuration
- 12mm opening diameter
- Polypropylene material
- Without handle
- Can be autoclaved at 121 °C
- Range of colors for each area of the laboratory
SARSTEDT's practical and versatile range of racks and storage boxes provides a solution for every application. The Block Rack D12 is designed for space-saving sample storage, and can be combined to form sample distribution blocks. This rack features 50 apertures for S-Monovettes and tubes with 8 to 12 mm diameter.
6 Options Available Below
Product Details & Documents
High-quality, safe, and robust racks and boxes are essential for specimen processing and storage. SARSTEDT offers the right product for every laboratory and analytical requirement.
- Format: 5×10 configuration
- 12mm opening diameter
- Polypropylene material
- Without handle
- Can be autoclaved at 121 °C
- Range of colors for each area of the laboratory
SARSTEDT's practical and versatile range of racks and storage boxes provides a solution for every application. The Block Rack D12 is designed for space-saving sample storage, and can be combined to form sample distribution blocks. This rack features 50 apertures for S-Monovettes and tubes with 8 to 12 mm diameter.
Product Family Options
Product Information
- ColorMaterialDimensionsAvailabilityPrice
- BluePP152×80×45 mmPack of 1$41.61Case of 20$832.20Supplier Number: 93.854.124
- GreenPP152×80×45 mmPack of 1$41.61Case of 20$832.20Supplier Number: 93.854.123
- GreyPP152×80×45 mmPack of 1$27.33Case of 20$546.60Supplier Number: 93.854
- OrangePP152×80×45 mmPack of 1$41.61Case of 20$832.20Supplier Number: 93.854.125
- RedPP152×80×45 mmPack of 1$41.61Case of 20$832.20Supplier Number: 93.854.121
- YellowPP152×80×45 mmPack of 1$41.61Case of 20$832.20Supplier Number: 93.854.122