Orders LinkContactUsLinkComponent
Color-Coded Inserts for CryoPure Tubes
Color-Coded Inserts for CryoPure Tubes
Supplier:   SARSTEDT INC
Color-Coded Inserts for CryoPure Tubes
Supplier:  SARSTEDT INC
Certificates
Color-Coded Inserts for CryoPure Tubes
Color-Coded Inserts for CryoPure Tubes
7 Options Available Below
Product Details & Documents
High-quality, safe and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Polystyrene material
  • Suitable for use with CryoPure tubes
  • Visual coding for easy identification of samples
  • Colors available individually or as a color mix
Up to 36 color combinations are available in combination with the screw caps in 6 different colors. Assorted color options include a mix of White, Red, Yellow, Green, and Purple.
Product Family Options
Product Information
  • Color
    Availability
    Price
  • Blue
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $6.13
    $6.13
    Log in for availability
    Case of 3000
    Case of 3000 - $183.90
    $183.90
    Pack of 100
       $6.13
    Case of 3000
       $183.90
    Supplier Number: 65.386.006
  • Green
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $6.13
    $6.13
    Log in for availability
    Case of 3000
    Case of 3000 - $183.90
    $183.90
    Pack of 100
       $6.13
    Case of 3000
       $183.90
    Supplier Number: 65.386.005
  • Purple
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $6.13
    $6.13
    Log in for availability
    Case of 3000
    Case of 3000 - $183.90
    $183.90
    Pack of 100
       $6.13
    Case of 3000
       $183.90
    Supplier Number: 65.386.007
  • Red
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $6.13
    $6.13
    Log in for availability
    Case of 3000
    Case of 3000 - $183.90
    $183.90
    Pack of 100
       $6.13
    Case of 3000
       $183.90
    Supplier Number: 65.386.002
  • White
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $6.13
    $6.13
    Log in for availability
    Case of 3000
    Case of 3000 - $183.90
    $183.90
    Pack of 100
       $6.13
    Case of 3000
       $183.90
    Supplier Number: 65.386
  • Yellow
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $6.13
    $6.13
    Log in for availability
    Case of 3000
    Case of 3000 - $183.90
    $183.90
    Pack of 100
       $6.13
    Case of 3000
       $183.90
    Supplier Number: 65.386.004
  • Assorted
    Log in for availability
    Log in for availability
    Case of 2,500
    Case of 2,500 - $208.01
    $208.01
    Case of 2,500
       $208.01
    Supplier Number: 65.386.992