Reaction Tubes with 'EASY CAP'
77588-748
New Product
- Cap type:Neutral, PP, 'EASY CAP'
- Volume:1.5 ml
- Color:Transparent
- Cat. No.:77588-748
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- Polypropylene retaining loop caps attached
- Polypropylene tube material
- Smoother stopper
- Larger packaging units for economical storage and stocking
- High centrifugation up to 20,000 ×g
This broad product portfolio in the field of test and micro tubes includes options with round, conical or flat base, with or without skirted base, with push cap or screw cap, with or without print or labels.