Lid for ELISA and Microtest Plates
Catalog # 77589-210
Supplier: SARSTEDT INC
New Product
Specifications
- Color:Transparent
- Description:Lid for ELISA and microtest plates
- Material:Polystyrene
- Cat. No.:77589-210
Specifications
About this item
Lids are suitable for use with ELISA and microtest plates in 96-well format.
- Transparent lid made from polystyrene
- Suitable for microtest and ELISA plates
- With condensation rings to reduce evaporation
These lids are free from DNA/DNase/RNase, free from pyrogens and endotoxins, and non-cytotoxic.