- Description:Steam-Dot™ Indicators
- Cat. No.:87001-938
- Supplier no.:364001
Steam-Dot™ indicators provide a fast positive visual check for processed packages sterilized in steam.
- Self-adhesive steam process indicator
- Permanent white to black color change on exposure to steam
- Decipher between processed and unprocessed packages
Steam-Dot™ Indicators offer a fast positive visual check for processed steam sterilized items. Steam-Dot™ Indicator changes from white to black after exposure to steam sterilization cycle.