Adjustable Gate Valve Lockout, Brady®
Catalog # 12222-798
Supplier: Brady Worldwide
Specifications
- Description:Adjustable Gate Valve Lockout
- Cat. No.:12222-798
- Supplier no.:64057
Specifications
About this item
Versatile, economical lockout adjusts to fit 2.5–16.5cm (1–61/2") diameter valve handles, replacing three conventional gate valve lockouts
Made of durable polypropylene for resistance to extreme temperature changes from –46 to 182°C (–50 to 360°F). Device is compact for easy carrying and storage and features front legend that reads: “DANGER. LOCKED OUT. DO NOT OPERATE.”