Orders LinkContactUsLinkComponent
Dual Position Ventilation Caps
Dual Position Ventilation Caps
Supplier:   SARSTEDT INC
Dual Position Ventilation Caps
Supplier:  SARSTEDT INC
Certificates
Dual Position Ventilation Caps
Dual Position Ventilation Caps
2 Options Available Below
Product Details & Documents
High-quality, safe, and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Two position fit allows both gas exchange and sealing
  • Ribbed surface for maximum grip
Dual position ventilation caps offer excellent sealing properties, and are available in LDPE material.
Product Family Options
Product Information
  • Material
    For tube O.D.
    Cap color
    Availability
    Price
  • 77588-226
    Dual position closure cap
    Dual position closure cap
    LDPE
    12 mm
    Neutral
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $6.24
    $6.24
    Log in for availability
    Case of 5000
    Case of 5000 - $312.00
    $312.00
    Pack of 100
       $6.24
    Case of 5000
       $312.00
    Supplier Number: 65.722
  • 77588-232
    Dual position closure cap
    Dual position closure cap
    LDPE
    16 mm
    Neutral
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $6.89
    $6.89
    Log in for availability
    Case of 5000
    Case of 5000 - $344.50
    $344.50
    Pack of 100
       $6.89
    Case of 5000
       $344.50
    Supplier Number: 65.725