Order Entry
United States
ContactUsLinkComponent
Tubes with Flat Base and Push Cap, PS
Tubes with Flat Base and Push Cap, PS
Catalog # 77587-674
Supplier:  SARSTEDT INC
New Product
Tubes with Flat Base and Push Cap, PS
Catalog # 77587-674
Supplier:  SARSTEDT INC
Supplier Number:  58.485.001
New Product

Specifications

  • Ø:
    16 mm
  • Sterility:
    Sterile
  • Volume:
    7 ml
  • Length:
    50 mm
  • Cat. No.:
    77587-674
  • Cap color:
    Neutral
  • Graduations:
    Non graduated

Specifications

About this item

High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.

  • Without graduations
  • Supplied sterile
  • Polystyrene (PS) tubes for maximum transparency
  • Ribbed cap for maximum grip
  • Flat base shape ensures the tube stands securely

This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.