Order Entry
United States
Orders LinkContactUsLinkComponent
Collection Cup, 125 ml

Collection Cup, 125 ml

125 ml, 85×62 mm (H×Dia.), Transparent
Catalog # 77589-066
Supplier:   SARSTEDT INC
Supplier #:  75.565
Collection Cup, 125 ml
Catalog # 77589-066
Supplier:  SARSTEDT INC
Collection Cup, 125 ml
Collection Cup, 125 ml

125 ml, 85×62 mm (H×Dia.), Transparent

Price
$158.61
Log in for availability
Case of 500
Product Details & Documents

High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.

  • Transparent polypropylene material
  • Break-proof and stackable
  • Graduated
  • Ideal height for test strips

SARSTEDT collection cups and containers are available in a range of sizes and volumes.

Specifications
  • Capacity:
    125 ml
  • Color:
    Transparent
  • Dimensions:
    85×62 mm (H×Dia.)