TNTplus™ Test Kits, COD
About this item
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process.
- Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
- Barcode helps the instrument to automatically detect and run the correct method
- Barcode allows spectrophotometer to detect reagent lot number and expiry date
- Vials have flat bottom to improve handling
- Vials are color coded by parameter and range
- No reagent blank necessary
4 Options Available Below
Product Details & Documents
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process.
- Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
- Barcode helps the instrument to automatically detect and run the correct method
- Barcode allows spectrophotometer to detect reagent lot number and expiry date
- Vials have flat bottom to improve handling
- Vials are color coded by parameter and range
- No reagent blank necessary
Product Family Options
Product Information
- RangeAvailabilityPrice
- Supplier Number: TNT820Specifications:MarketsourceMarketsource
- Supplier Number: TNT821Specifications:MarketsourceMarketsource
- Supplier Number: TNT82206Specifications:MarketsourceMarketsource
- Supplier Number: TNT82306Specifications:MarketsourceMarketsource