Order Entry
United States
ContactUsLinkComponent
Silver Nitrate Standard Solution
Silver Nitrate Standard Solution
  104085-260
 :  Hach
 :  
undefined
Silver Nitrate Standard Solution
  104085-260
 :  Hach
 :  1255149
 :  

 

  • Description:
    Silver nitrate standard solution, 0.141 N, 500 ml
  • Size:
    500 ml
  • Cat. No.:
    104085-260

 

 

Standard solution for chloride determination by argentometric burette titration.