Multi-Parameter Meters, Bench, HQd (HQ430d / HQ440d)
Supplier: Hach
Certificates
About this item
These advanced laboratory meters with one or two input channels are ideal for testing critical water quality parameters.
- Single- or dual-input channel for pH, ORP, mV, temperature, conductivity, TDS, salinity, DO and ISE: For use with any IntelliCAL™ electrode
- Simple user interface that does not require manuals or training to operate
- Guided calibration and check standard routines reduce calibration errors while stabilization alerts
- Internal USB ports simplify data transfer, peripherals and software updates, and connection to PCs printer and keyboards
- Fully GLP compliant communication and documentation
- Ultra-bright screen and large font size for easy to read results
The meters connect with smart probes (IntelliCAL™) that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and set-up time. They are designed to withstand years of continuous lab use.
Ordering information: Supplied with four batteries, universal power adapter, USB cable and electrode holder. IntelliCAL™ electrodes need to be ordered separately.
Specifications
- Catalog No:
- 104087-306
- 104087-308
- pH Range:
- 0 - 14
- 0 - 14
- pH Resolution:
- 0.1/ 0.01/ 0.001
- 0.1/ 0.01/ 0.001
- pH Accuracy:
- ±0.002
- ±0.002
- pH Calibration:
- Color-coded: 4.01/7.00/10.01; IUPAC: 1.679/4.005/7.000/10.012/12.45; DIN: 1.09/4.65/9.23; user-defined custom buffer sets
- Color-coded: 4.01/7.00/10.01; IUPAC: 1.679/4.005/7.000/10.012/12.45; DIN: 1.09/4.65/9.23; user-defined custom buffer sets
- mV range:
- ±1500
- ±1500
- mV resolution:
- 0.1/ 0.01/ 0.001
- 0.1/ 0.01/ 0.001
- mV accuracy:
- ±0.1
- ±0.1
- ISE Range:
- Electrode specific
- Electrode specific
- ISE Resolution:
- 0.1/0.01/0.001
- 0.1/0.01/0.001
- ISE Accuracy:
- ±0.1 mV
- ±0.1 mV
- Conductivity Range:
- 0.01 μS/cm - 200 mS/cm
- 0.01 μS/cm - 200 mS/cm
- Conductivity Resolution:
- 5 digits with 2 digits after decimal point
- 5 digits with 2 digits after decimal point
- Conductivity Accuracy:
- ±0.5% (1 μS/cm - 200 mS/cm)
- ±0.5% (1 μS/cm - 200 mS/cm)
- Conductivity Calibration:
- Demal (1D / 0.1D / 0.01D); Molar (0.1M / 0.01M / 0.001M); NaCl (0.05%/25 µS/cm/1000 µS/cm/18 mS/cm); standard sea water; user defined
- Demal (1D / 0.1D / 0.01D); Molar (0.1M / 0.01M / 0.001M); NaCl (0.05%/25 µS/cm/1000 µS/cm/18 mS/cm); standard sea water; user defined
- Resistivity range:
- 2.5 Ωcm - 49 MΩcm
- 2.5 Ωcm - 49 MΩcm
- Resistivity accuracy:
- ±0.5%
- ±0.5%
- Salinity Range:
- 0 - 42 g/kg
- 0 - 42 g/kg
- Salinity Resolution:
- 0.01 ppt
- 0.01 ppt
- Salinity Accuracy:
- ±0.1 mg/L (for range <8 mg/L)
- ±0.1 mg/L (for range <8 mg/L)
- TDS Range:
- 0.0 - 50.0 g/L
- 0.0 - 50.0 g/L
- TDS Resolution:
- max. 3 digits
- max. 3 digits
- TDS Accuracy:
- ±0.5%
- ±0.5%
- DO Concentration:
- 0.00 - 20.0 mg/L
- 0.00 - 20.0 mg/L
- DO Resolution:
- 0.01 mg/L
- 0.01 mg/L
- Temperature Resolution:
- 0.1 °C
- 0.1 °C
- Temperature Accuracy:
- ±0.3 °C
- ±0.3 °C
- Temperature Compensation:
- Automatic, off, manual
- Automatic, off, manual
- Display:
- Backlit graphical LCD
- Backlit graphical LCD (with simultaneous readings from two electrodes)
- Power supply:
- 230 V with power supply or 4 × AA Alkaline batteries
- 230 V with power supply or 4 × AA Alkaline batteries
- Data Logging Capacity:
- 500 results
- 500 results
- Connections:
- 1 × M12 digital for IntelliCAL™ electrodes
- 2 × M12 digital for IntelliCAL™ electrodes
- Outputs:
- USB
- USB
- W×D×H:
- 175×234×86 mm
- 175×234×86 mm
- Weight:
- 0.85 kg
- 0.85 kg
Specifications
Frequently Bought Together