Order Entry
United States
ContactUsLinkComponent
 
pH/mV/°C Meter, Bench, HQd (HQ411D)
Catalog # 104087-304
Supplier:  Hach
undefined
pH/mV/°C Meter, Bench, HQd (HQ411D)
Catalog # 104087-304
Supplier:  Hach
Supplier Number:  HQ411D
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Temperature Accuracy:
    ±0.3 °C
  • Temperature Resolution:
    0.1 °C
  • Automatic Buffer Recognition:
    Color-coded: 4.01, 7.00, 10.01 pH;
    IUPAC: 1.679, 4.005, 7.000, 10.012, 12.45 pH
    DIN: 1.09, 4.65, 9.23
    User-defined custom buffer sets
  • Connections:
    1× M12 digital for IntelliCAL™ electrodes
  • Data Logging Capacity:
    500 results
  • Display:
    Backlit graphical LCD
  • Outputs:
    USB
  • Power supply:
    230 V with power supply or 4× AA Alkaline batteries
  • Temperature Compensation:
    Automatic, off, manual
  • Weight:
    0.85 kg
  • W×D×H:
    175×234×86 mm
  • mV accuracy:
    ±0.1
  • mV range:
    ±1500
  • mV resolution:
    0.1/ 0.01/ 0.001
  • pH Accuracy:
    ±0.002
  • pH Range:
    0 - 14
  • pH Resolution:
    0.1/ 0.01/ 0.001
  • Cat. No.:
    104087-304

Specifications

About this item

This advanced laboratory meter is ideal for testing critical water quality parameters. The meter connects with smart probes (IntelliCAL™) that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The instrument is designed to withstand years of continuous lab use.

  • Single input channel for pH, ORP, mV, and temperature: For use with any IntelliCAL™ smart pH or ORP electrode
  • Simple user interface that does not require manuals or training to operate
  • Guided calibration and check standard routines reduce calibration errors
  • Internal USB ports simplify data transfer, peripherals, and software updates and connection to PCs printer and keyboards
  • Fully GLP compliant communication and documentation
  • Ultra-bright screen and large font size for easy to read results
Ordering information: Supplied with four batteries, universal power adapter and with or without electrode holder. IntelliCAL™ electrodes need to be ordered separately.