Order Entry
United States
Orders LinkContactUsLinkComponent
Buffer Solution for Chlorine Determination

Buffer Solution for Chlorine Determination

Marketsource
Supplier:   Hach
Buffer Solution for Chlorine Determination
Supplier:  Hach
Marketsource
Certificates
Buffer Solution for Chlorine Determination
Buffer Solution for Chlorine Determination

2 Options Available Below

Product Details & Documents

For determination of chlorine by amperometric titration. APHA formulation 100 ml bottle.

Product Family Options

Product Information

  • pH buffer type
    pH
    Availability
    Price
  • Acetate
    4
    Log in for availability
    Log in for availability
    Each
    Each - $46.09
    $46.09
    Each
       $46.09
    Minimum order quantity is 1

    Supplier Number: 1490932
    Specifications:
    Marketsource
    Marketsource
  • Phosphate
    7
    Log in for availability
    Log in for availability
    Each
    Each - $43.46
    $43.46
    Each
       $43.46
    Minimum order quantity is 1

    Supplier Number: 2155332
    Specifications:
    Marketsource
    Marketsource