ContactUsLinkComponent
Soil Extractant Solution, Mehlich 2
Soil Extractant Solution, Mehlich 2
Catalog # 104086-016
Supplier:  Hach
CAS Number:  
undefined
Soil Extractant Solution, Mehlich 2
Catalog # 104086-016
Supplier:  Hach
Supplier Number:  2266349
CAS Number:  
Specifications
  • Description:
    Mehlich 2 soil extractant solution
  • Size:
    500 ml
  • Cat. No.:
    104086-016
Specifications
About this item
The Mehlich 2 extractant is used for phosphate and potassium determination in soils.