Order Entry
United States
Orders LinkContactUsLinkComponent
pH Electrode Filling Solution

pH Electrode Filling Solution

pH electrode filling solution, 4M KCl saturated with silver chloride (AgCl ), 65 ml, 65 ml
Catalog # 104086-866
Marketsource
Supplier:   Hach
Supplier #:  5194100
pH Electrode Filling Solution
Catalog # 104086-866
Supplier:  Hach
Marketsource
CAS Number:  
pH Electrode Filling Solution

pH electrode filling solution, 4M KCl saturated with silver chloride (AgCl ), 65 ml, 65 ml

CAS Number:  
Price
$32.11
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Specifications

Specifications

  • Description:
    pH electrode filling solution, 4M KCl saturated with silver chloride (AgCl ), 65 ml
  • Size:
    65 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.