ContactUsLinkComponent
Visual Colour Kits, Phosphorus, PO-19A
Visual Colour Kits, Phosphorus, PO-19A
Catalog # 104086-000
Supplier:  Hach
CAS Number:  
undefined
Visual Colour Kits, Phosphorus, PO-19A
Catalog # 104086-000
Supplier:  Hach
Supplier Number:  224801
CAS Number:  
Specifications
  • Cat. No.:
    104086-000
  • Range:
    0 - 0.8 mg/L PO₄
    0 - 4 mg/L PO₄
    0 - 40 mg/L PO₄
Specifications
About this item
Simple, economical rapid color disc method with an easy to carry test kit.
Delivery: Kit includes two reagents, color disc, comparator box, two viewing tubes, viewing adaptor, two bottles, dropper tube, funnel, filter paper, deionized water, instructions and carrying case.