Orders LinkContactUsLinkComponent
Multi Parameter Test Kit
Multi Parameter Test Kit
Multi parameter test kit, CN-39WR
Catalog # 104085-950
Marketsource
Supplier:   Hach
Supplier #:  223002
Multi Parameter Test Kit
Catalog # 104085-950
Supplier:  Hach
Marketsource
CAS Number:  
Multi Parameter Test Kit
Multi Parameter Test Kit
Multi parameter test kit, CN-39WR
CAS Number:  
Price
$555.23
Log in for availability
Pack of 100
Minimum order quantity is 1
Product Details & Documents
This multi-test kit combines popular water treatment tests into one compact kit which can be used to determine chlorine (0 to 3.4 mg/L), hardness (1 to 20 gpg), iron (0 to 4 mg/L) and pH (4 to 10) in the water sample.
  • Includes frequently measured water quality parameters
  • Uses simple, economical methods
Hardeness testing: gpg= grains per gallon
Delivery information: Includes six reagents, bottles, colour discs, comparator box, viewing tubes, measuring tube, instructions and carrying case.
Specifications
Specifications
  • Description:
    Multi parameter test kit, CN-39WR