ContactUsLinkComponent
Nitrites Test Kits, NI-15
Catalog # 104085-892
Supplier:  Hach
CAS Number:  
undefined
Nitrites Test Kits, NI-15
Catalog # 104085-892
Supplier:  Hach
Supplier Number:  2182000
CAS Number:  
Specifications
  • Cat. No.:
    104085-892
  • Range:
    0 - 0.40 mg/L NO₂- N
Specifications
About this item
The accurate visual test kit method to determine nitrogen as nitrite by diazotisation of sample followed by color comparison.
  • Simple and cost effective
  • Portable test kit
Delivery: Kit includes color disc, comparator box and tubes, reagents (but not deionized water), instructions and carrying case.