ContactUsLinkComponent
Nitrite Test Kit, Colour Disc, NI-6
Nitrite Test Kit, Colour Disc, NI-6
Catalog # 104085-964
Supplier:  Hach
CAS Number:  
undefined
Nitrite Test Kit, Colour Disc, NI-6
Catalog # 104085-964
Supplier:  Hach
Supplier Number:  224000
CAS Number:  
Specifications
  • Cat. No.:
    104085-964
  • Range:
    0 - 80 mg/L NO₂
    0 - 1600 mg/L NO₂
Specifications
About this item
The kit uses the color disc or ferrous sulphate method to measure nitrogen as nitrite NO²⁻.
  • Simple, cost effective
  • Portable test kit for analysis in the field
Delivery: Kit includes color disc, comparator box and tubes, reagents, deionized dilution water, instructions and carrying case.