Orders LinkContactUsLinkComponent
DeColorizer, pPb-1
DeColorizer, pPb-1
pPb-6, decolorizer, 10 ml, 10 ml, LeadTrak™
  104086-170
Marketsource
 :   Hach
Supplier #:  2374755
DeColorizer, pPb-1
  104086-170
 :  Hach
Marketsource
 :  
DeColorizer, pPb-1
DeColorizer, pPb-1
pPb-6, decolorizer, 10 ml, 10 ml, LeadTrak™
 :  
$57.66
Each
Minimum order quantity is 1
They are used for determining lead by the LeadTrak™ Fast Column Extraction method.
Documents
  • Chemical method:
    LeadTrak™
  • Description:
    pPb-6, decolorizer, 10 ml
  • Size:
    10 ml