Orders LinkContactUsLinkComponent
Media set
Media set
Total bacteria media set, for total count, 3 tests
Catalog # 104086-042
Marketsource
Supplier:   Hach
Supplier #:  2277700
Media set
Catalog # 104086-042
Supplier:  Hach
Marketsource
CAS Number:  
Media set
Media set
Total bacteria media set, for total count, 3 tests
CAS Number:  
Price
$87.09
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Media set is a colorimetric method that is used for determination of total bacteria.
  • ToxTrak™ Toxicity test speeds sample evaluations
  • Economically alternative to bioassays
It includes dehydrated tryptic soy broth, screw cap vial and sterile transfer pippets.
Specifications
Specifications
  • Description:
    Total bacteria media set, for total count
  • Size:
    3 tests
Specifications