ContactUsLinkComponent
ECR Masking Reagent Solution
ECR Masking Reagent Solution
Catalog # 104086-176
Supplier:  Hach
CAS Number:  
undefined
ECR Masking Reagent Solution
Catalog # 104086-176
Supplier:  Hach
Supplier Number:  2380123
CAS Number:  
Specifications
  • Cat. No.:
    104086-176
  • Range:
    0.006 - 0.25 mg/L
Specifications
About this item
This solution is used for Aluminum determination by the Eriochrome Cyanine R Method.