ContactUsLinkComponent
Stablcal® Turbidity Standard, ULR Verification Kit
Stablcal® Turbidity Standard, ULR Verification Kit
Catalog # 104086-596
Supplier:  Hach
CAS Number:  
undefined
Stablcal® Turbidity Standard, ULR Verification Kit
Catalog # 104086-596
Supplier:  Hach
Supplier Number:  2714600
CAS Number:  
Specifications
  • Turbidity:
    0.10 NTU
    0.50 NTU
    1.0 NTU
    0.30 NTU
  • Description:
    ULR verification kit, Stablcal®
  • Size:
    4× 100 ml
  • Cat. No.:
    104086-596
Specifications
About this item
Stablcal® verification kit containing 0.10 NTU, 0.30 NTU, 0.50 NTU, 1.0 NTU standards (100 ml each), sililcone oil, oiling cloth and instructions is used for verification purpose only.
  • Precise, pre-mixed formazin solutions
  • Enhanced stability saves time and labor
  • Confidence in results