ContactUsLinkComponent
Water Quality Test Discs, HA-77
Water Quality Test Discs, HA-77
Catalog # 104085-694
Supplier:  Hach
CAS Number:  
undefined
Water Quality Test Discs, HA-77
Catalog # 104085-694
Supplier:  Hach
Supplier Number:  202300
CAS Number:  
Specifications
  • Cat. No.:
    104085-694
  • Range:
    0 - 5 mg/L
Specifications
About this item
Hardness and iron determination in a water quality test kit which offers accuracy and portable utility in one package. Utilising drop count titration (hardness) and color disc colorimetry.
  • Easy to use
  • Portable kit
Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Delivery: Includes four reagents, bottles, flask, color discs, comparator box, viewing tubes, instructions and carrying case sufficient for 100 tests.