ContactUsLinkComponent
Reagent Powder Pillows for Determination of Molybdenum, HS-C
Reagent Powder Pillows for Determination of Molybdenum, HS-C
Catalog # 104086-388
Supplier:  Hach
CAS Number:  
undefined
Reagent Powder Pillows for Determination of Molybdenum, HS-C
Catalog # 104086-388
Supplier:  Hach
Supplier Number:  2537800
CAS Number:  
Specifications
  • Cat. No.:
    104086-388
  • Range:
    0 - 5 mg/L H₂S
Specifications
About this item
The kit is used to determine sulphide pollution of sewage water, waste water and other sources resulting from decomposition of organic matter. In the HS-C kit, sulphide discolours a detection paper. The results can be obtained by comparison of the color to a printed chart.
  • Reliable, cost effective color comparison method
Delivery: Includes reaction vial, effervescent tablets, test papers, comparator chart and instructions, in a cardboard box.