ContactUsLinkComponent
Sulphate Test Kit, SF-1
Sulphate Test Kit, SF-1
Catalog # 104086-006
Supplier:  Hach
CAS Number:  
undefined
Sulphate Test Kit, SF-1
Catalog # 104086-006
Supplier:  Hach
Supplier Number:  225100
CAS Number:  
Specifications
  • Cat. No.:
    104086-006
  • Range:
    50 - 200 mg/L
Specifications
About this item
High concentrations of magnesium or sodium sulphate in water act as cathartics. This test kit provides economical determination of sulphate levels using the Extinction/Turbidimetric methods.
Delivery: Includes graduated cylinder, mixing bottle, reagent, dipstick comparator, instructions and carrying case.