Orders LinkContactUsLinkComponent
pH Electrode Storage Solution
pH electrode storage solution, powder pillows (each pillow makes 25 ml of storage solution)
  104086-508
Marketsource
 :   Hach
 :  2657364
pH Electrode Storage Solution
  104086-508
 :  Hach
Marketsource
 :  
pH Electrode Storage Solution
pH Electrode Storage Solution
pH electrode storage solution, powder pillows (each pillow makes 25 ml of storage solution)
 :  
$83.00
Pack of 20
  • Description:
    pH electrode storage solution, powder pillows (each pillow makes 25 ml of storage solution)