Chloride Standard Solution
104085-632
: Hach
:
- Concentration:1000 mg/L
- Description:Chloride standard solution
- Size:500 ml
- Cat. No.:104085-632
For accuracy checks in chloride determinations. Also suitable for standardising conductivity apparatus.