Fluoride Standard Solution
Catalog # 104086-670
Supplier: Hach
CAS Number:
Specifications
- Concentration:5 mg/l as F
- Description:Fluoride standard solution
- Size:500 ml
- Cat. No.:104086-670
Specifications
About this item
For accuracy checks in fluoride determinations.