Orders LinkContactUsLinkComponent
pH Indicator Strips
pH Indicator Strips
1, 0 - 14.0
Catalog # 104086-454
Marketsource
Supplier:   Hach
Supplier #:  2601300
pH Indicator Strips
Catalog # 104086-454
Supplier:  Hach
Marketsource
pH Indicator Strips
pH Indicator Strips
1, 0 - 14.0
Price
$61.05
Log in for availability
Pack of 100
Minimum order quantity is 1
Product Details & Documents
pH strips provides a simple, fast and economical method of pH measurement. Can obtain accurate results in weakly buffered, turbid and lightly colored samples. It is easy to use and disposable.
Specifications
  • pH range:
    0 - 14.0
  • pH graduation:
    1