ContactUsLinkComponent
MEL/m-ColiBlue24® Field Filtration Kit
MEL/m-ColiBlue24® Field Filtration Kit
  104086-432
 :  Hach
 :  
undefined
MEL/m-ColiBlue24® Field Filtration Kit
  104086-432
 :  Hach
 :  2586400
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Field filtration kit, MEL/m-ColiBlue24®
  • Environmentally Preferable:
  • Cat. No.:
    104086-432
The coliform and E.coli field filtration kit features m-ColiBlue24® broth and Millipore's portable filter assembly, push fit funnels and Millipore membranes to simplify membrane filtration testing in the field and also m-ColiBlue24® broth simultaneously detects total coliforms and E.coli in 24 hours.
  • Efficient field operations
  • Includes consumables and media
  • EPA approved field method
 : The carrying case contains 40 m-ColiBLue24® broth ampules, 150 push fit funnels and S-Pak filters (Millipore), 1 microfil stand alone support (Millipore), 1 syringe/tubing vacuum device, 50 petri dishes (50 mm, sterile) with nutrient pad, 50 sterile, individually wrapped 10 ml pipets (poly), 50 sterile sample bags with dechlorinating agent, alcohol burner and 100 ml of isopropyl alcohol, illuminated magnifier (5X), pipette filler, forceps, ampule breaker and procedure documentation.