SHOWA CS701 Chemical Resistant Gloves
: Showa
New Product
The CS701 food-safe gloves are designed with precision in mind. Anti-slip, rough surface grip gives the wearer ice-grip and optimum performance.
- Anti-slip grip
- Chemical-resistant; Oil-resistant
- Seamless knit
- Robust durability
- Forearm protection
- Natural rubber latex-free
- Double coated Nitrile
- Polyester liner; Rough grip
A double-dipped nitrile coating also provides chemical protection and durability to ensure safety from dangerous substances.
The seamless liner and natural latex-free composition are skin-friendly and comfortable for long-wear.
Applications:
- Chemical spray and treatment
- Light chemical handling in horticulture, agriculture
- Coating preparation
- Laboratory, pharma and analysis
- Painting and spray workshops
- Fruit and vegetable processing
- Grain, mill and starch products
- Washing and cleaning
: CE cat. III, EN 388:2016+A1:2018, EN ISO 374-1:2016+A1:2018/Type A, EN ISO 374-5:2016, EN 407:2004 certified.
: 10 pair per polybag, 6 polybags per case, 60 pair per case.
- Catalog No:
- 76521-136
- 76521-140
- 76521-138
- 76521-132
- 76521-134
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable/Reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Material:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Food Contact Approved:
- +
- +
- +
- +
- +
Frequently Bought Together