Order Entry
United States
ContactUsLinkComponent
Trypsin-EDTA (0.25%)
Trypsin-EDTA (0.25%)
Catalog # MSPP-07901
CAS Number:  
undefined
Trypsin-EDTA (0.25%)
Catalog # MSPP-07901
Supplier Number:  07901
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Species:
    Human
    Mouse
    Non-human primate
    Rat
  • Description:
    Trypsin-EDTA (0.25%)
  • Size:
    500 ml
  • Cat. No.:
    MSPP-07901

Specifications

About this item

Enzymatic cell dissociation reagent.

  • Dissociate cells efficiently with a combined solution
  • EDTA enhances Trypsin-mediated cell detachment

Use Trypsin-EDTA (0.25%) for enzymatic dissociation of adherent cells, cell aggregates and tissues into single-cell suspensions. EDTA weakens cell-cell adhesion, which improves trypsin's access to hydrolysis-targeted peptide bonds, and enhances detachment of adherent cells. Trypsin-EDTA is also available in a lower concentration (0.05%), for applications that require more gentle dissociation.

Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').