
Potassium hydroxide 8 mol/L (8 N) in aqueous solution
Catalog # 104086-696
Supplier: Hach
CAS Number:
Specifications
- Cat. No.:104086-696
Specifications
About this item
:56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,III
:GHS05
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553