Order Entry
United States
ContactUsLinkComponent
Sodium hydroxide 5 mol/L (5 N) in aqueous solution
Sodium hydroxide 5 mol/L (5 N) in aqueous solution
Catalog # 104086-310
Supplier:  Hach
CAS Number:  
Sodium hydroxide 5 mol/L (5 N) in aqueous solution
Catalog # 104086-310
Supplier:  Hach
Supplier Number:  245026
CAS Number:  

Specifications

  • Cat. No.:
    104086-310

Specifications

About this item

 :40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:≤1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
 :
Formula:NaOH
MDL Number:MFCD00003548